siRNA Details
Gene Symbol: Triobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024716.1 | NP_001019887.1 | 2 | 190 | ||
NM_001039156.1 | NP_001034245.1 | 13 | 4957 | |||
NM_138579.4 | NP_613045.3 | 12 | 4819 | |||
GenBank mRNA | AK053194.1 | - | 2 | 150 | ||
AK085512.1 | - | 2 | 160 | |||
AK129417.1 | - | 2 | 140 | |||
AK155197.1 | - | 5 | 2522 | |||
AK170390.1 | - | 2 | 136 | |||
AK197781.1 | - | 2 | 81 | |||
BC042760.1 | - | 2 | 129 | |||
BC053329.1 | - | 2 | 174 | |||
BY199415.1 | - | 2 | 191 | |||
CF533172.1 | - | 2 | 262 | |||
DQ228000.1 | - | 13 | 4957 | |||
DQ228001.2 | - | 12 | 4819 | |||
DQ278602.1 | - | 11 | 4438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
