siRNA Details
Gene Symbol: Triobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039155.1 | NP_001034244.1 | 8 | 3751 | ||
GenBank mRNA | BC094366.1 | - | 1 | 2406 | ||
DQ228002.2 | - | 8 | 3751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
