siRNA Details
Gene Symbol: Syne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079686.1 | NP_001073154.1 | 55 | 9357 | ||
NM_022027.2 | NP_071310.2 | 17 | 3311 | |||
GenBank mRNA | AK036828.1 | - | 18 | 3367 | ||
BC041779.1 | - | 16 | 2908 | |||
BC054456.1 | - | 17 | 3311 | |||
BC110426.1 | - | 25 | 4559 | |||
BC110428.1 | - | 17 | 3291 | |||
BC111034.1 | - | 6 | 1107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
