siRNA Details
Gene Symbol: Pde4dip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178080.4 | NP_835181.2 | 17 | 3944 | ||
GenBank mRNA | AK028810.1 | - | 17 | 3911 | ||
AK122286.1 | - | 17 | 3939 | |||
BC033477.1 | - | 1 | 677 | |||
BC034266.1 | - | 17 | 2609 | |||
BC050783.1 | - | 17 | 3898 | |||
BC057350.1 | - | 17 | 4375 | |||
BC141172.1 | - | 17 | 3872 |
