siRNA Details
Gene Symbol: Gm10220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134299.1 | NP_001127771.1 | 2 | 247 |
Gene Symbol: 5031410I06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207657.3 | NP_997540.2 | 2 | 195 |
