siRNA Details
Gene Symbol: Flad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110138.1 | NP_001103608.1 | 3 | 309 | ||
GenBank mRNA | BC161933.1 | - | 3 | 328 | 9022122 | |
FQ218677.1 | - | 3 | 339 |

More Information
Other Species Targeted

Gene Ontology Categories:
