siRNA Details
Gene Symbol: Ube2e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108371.1 | NP_001101841.1 | 3 | 343 | ||
XM_006251699.2 | XP_006251761.2 | 4 | 1148 |

More Information
Other Species Targeted

Gene Ontology Categories:
