siRNA Details
Gene Symbol: Tsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115027.1 | NP_001108499.1 | 4, 5 | 468 | ||
XM_006256781.2 | XP_006256843.1 | 3, 4 | 977 | |||
GenBank mRNA | BC158694.1 | - | 4, 5 | 467 | 9023223 | |
FQ216460.1 | - | 4, 5 | 458 |
Gene Symbol: LOC100910723 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752029.3 | XP_003752077.2 | 719 |

More Information
Other Species Targeted

Gene Ontology Categories:
