siRNA Details
Gene Symbol: Zfp786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127594.1 | NP_001121066.1 | 4 | 1750 | ||
XM_006236413.2 | XP_006236475.1 | 4 | 1807 | |||
XM_006236414.2 | XP_006236476.1 | 4 | 1893 | |||
GenBank mRNA | BC166479.1 | - | 4 | 1750 | 9102251 |
