siRNA Details
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039028.2 | NP_001034117.2 | 4 | 382 | ||
GenBank mRNA | BC166803.1 | - | 4 | 349 | 7938869 | |
EV779654.1 | - | 4 | 410 | |||
FQ228979.1 | - | 4 | 350 |

More Information
Other Species Targeted

Gene Ontology Categories:
