siRNA Details
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039028.2 | NP_001034117.2 | 5 | 486 | ||
GenBank mRNA | BC166803.1 | - | 5 | 453 | 7938869 | |
DY315273.1 | - | 2 | 99 | |||
EV779654.1 | - | 5 | 514 | |||
FQ228979.1 | - | 5 | 454 |
