siRNA Details
Gene Symbol: Rpgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773079.1 | XP_008771301.1 | 10 | 1497 | ||
XM_008773081.1 | XP_008771303.1 | 10 | 1499 | |||
XM_008773082.1 | XP_008771304.1 | 10 | 1498 | |||
XM_008773083.1 | XP_008771305.1 | 10 | 1491 | |||
GenBank mRNA | BC162028.1 | - | 10 | 1235 | 9100258 |
Gene Symbol: LOC302586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773113.1 | XP_008771335.1 | 1 | 1228 |
