siRNA Details
Gene Symbol: Defa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025763.1 | NP_001020934.1 | 2 | 186 | ||
GenBank mRNA | AY623753.1 | - | 2 | 186 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Defa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025763.1 | NP_001020934.1 | 2 | 186 | ||
GenBank mRNA | AY623753.1 | - | 2 | 186 |