siRNA Details
Gene Symbol: Ndufv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101011.2 | NP_001094481.1 | 2 | 206 | ||
NM_022607.2 | NP_072129.2 | 2 | 206 | |||
GenBank mRNA | AB000098.1 | - | 2 | 137 | ||
BC059134.1 | - | 162 | ||||
CK365764.1 | - | 2 | 217 | |||
DV725422.1 | - | 2 | 204 | |||
FQ216920.1 | - | 2 | 154 | |||
FQ217920.1 | - | 2 | 154 | |||
FQ218041.1 | - | 2 | 154 | |||
FQ223657.1 | - | 2 | 154 |
