siRNA Details
Gene Symbol: Pnrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103360.1 | NP_001096830.1 | 3 | 717 | ||
GenBank mRNA | BC081903.1 | - | 3 | 717 | 7110880 |

More Information
Other Species Targeted

Gene Ontology Categories:
