siRNA Details
Gene Symbol: Mpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107036.1 | NP_001100506.1 | 7 | 1367 | ||
XM_006247095.2 | XP_006247157.1 | 12 | 1824 | |||
XM_006247096.2 | XP_006247158.1 | 12 | 1826 |
