siRNA Details
Gene Symbol: Hras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098241.1 | NP_001091711.1 | 3 | 319 | ||
NM_001130441.1 | NP_001123913.1 | 3 | 319 | |||
XM_006230535.2 | XP_006230597.2 | 3 | 313 | |||
GenBank mRNA | BC086608.1 | - | 3 | 315 | ||
BC099130.1 | - | 3 | 341 | |||
CO567790.1 | - | 3 | 319 |
