siRNA Details
Gene Symbol: Zbtb7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127375.1 | NP_001120847.1 | 5 | 1452 | ||
GenBank mRNA | BC166476.1 | - | 5 | 1452 | 9101314 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145356, AK045522, AK156372, BC018187, BC070424.
Gene Ontology Categories:
