siRNA Details
Gene Symbol: Zbtb7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127375.1 | NP_001120847.1 | 5 | 1519 | ||
GenBank mRNA | BC166476.1 | - | 5 | 1519 | 9101314 |

More Information
Other Species Targeted

Gene Ontology Categories:
