siRNA Details
Gene Symbol: RGD1565959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106886.1 | NP_001100356.1 | 2 | 289 | ||
XM_008766844.1 | XP_008765066.1 | 2 | 198 | |||
XM_008766845.1 | XP_008765067.1 | 2 | 198 |
