siRNA Details
Gene Symbol: Vps13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100975.2 | NP_001094445.2 | 48 | 6732 | ||
XM_008760289.1 | XP_008758511.1 | 49 | 7031 | |||
GenBank mRNA | AY325251.1 | - | 3 | 522 |
