siRNA Details
Gene Symbol: Adh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126120.1 | NP_001119592.1 | 8 | 1039 | ||
GenBank mRNA | BC083724.1 | - | 8 | 1039 | ||
FQ212950.1 | - | 8 | 1095 | |||
FQ213560.1 | - | 8 | 1085 |

More Information
Other Species Targeted

Gene Ontology Categories:
