siRNA Details
Gene Symbol: LOC100125362 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103354.2 | NP_001096824.1 | 2 | 669 | ||
XM_006243064.2 | XP_006243126.1 | 2 | 959 | |||
XM_008766199.1 | XP_008764421.1 | 2 | 819 | |||
GenBank mRNA | BC092621.1 | - | 1 | 515 | 7313228 |
