siRNA Details
Gene Symbol: Mpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021265.3 | NP_067088.2 | 20 | 1684 | ||
XM_006245011.2 | XP_006245073.1 | 21 | 2971 | |||
XM_008767130.1 | XP_008765352.1 | 20 | 2950 | |||
XM_008767131.1 | XP_008765353.1 | 22 | 2111 | |||
XM_008767132.1 | XP_008765354.1 | 23 | 1997 | |||
XM_008767133.1 | XP_008765355.1 | 19 | 1566 | |||
GenBank mRNA | AB030499.1 | - | 14 | 1501 | ||
AB030500.1 | - | 15 | 1555 | |||
AB030501.1 | - | 14 | 1447 |
