siRNA Details
Gene Symbol: Bnipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128187.2 | NP_001121659.1 | 6, 7 | 765 | ||
XM_006232889.2 | XP_006232951.1 | 5, 6 | 1104 | |||
GenBank mRNA | BC166747.1 | - | 6, 7 | 765 | 5598850 |

More Information
Other Species Targeted

Gene Ontology Categories:
