siRNA Details
Gene Symbol: Ubr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234339.1 | XP_006234401.1 | 1 | 591 | ||
XM_008761950.1 | XP_008760172.1 | 1 | 592 | |||
XM_008761951.1 | XP_008760173.1 | 1 | 592 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177783, NM_001081548, BC145475, DQ924536, AK037622.
Gene Ontology Categories:
