siRNA Details
Gene Symbol: RGD1305587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127452.1 | NP_001120924.1 | 370 | |||
GenBank mRNA | BC158618.1 | - | 370 | 7389133 |
Gene Symbol: LOC685881 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239781.2 | XP_006239843.1 | 1 | 167 |
