siRNA Details
Gene Symbol: Vangl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109584.1 | NP_001103054.1 | 4 | 506 | ||
XM_006233054.2 | XP_006233116.1 | 4 | 547 | |||
XM_006233055.2 | XP_006233117.1 | 4 | 546 |

More Information
Other Species Targeted

Gene Ontology Categories:
