siRNA Details
Gene Symbol: Eefsec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109249.1 | NP_001102719.1 | 2 | 344 | ||
XM_006236864.2 | XP_006236926.1 | 2 | 358 | |||
XM_008763079.1 | XP_008761301.1 | 2 | 359 | |||
GenBank mRNA | BC168694.1 | - | 2 | 321 | 7937199 |

More Information
Other Species Targeted

Gene Ontology Categories:
