siRNA Details
Gene Symbol: Ppcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039010.2 | NP_001034099.2 | 3 | 868 | ||
XM_008763998.1 | XP_008762220.1 | 3 | 653 | |||
XM_008763999.1 | XP_008762221.1 | 3 | 438 | |||
XM_008764000.1 | XP_008762222.1 | 2 | 576 | |||
GenBank mRNA | BC099208.1 | - | 2 | 1636 | 7384489 |
