siRNA Details
Gene Symbol: Hoxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107042.1 | NP_001100512.1 | 1 | 868 | ||
XM_006247194.2 | XP_006247256.1 | 3 | 765 | |||
GenBank mRNA | BC166859.1 | - | 3 | 765 | 9100655 |
Gene Symbol: Hoxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257706.2 | XP_006257768.1 | 4 | 6525 | ||
XM_006257707.2 | XP_006257769.1 | 2 | 822 | |||
XM_008773712.1 | XP_008771934.1 | 3 | 4555 |
Gene Symbol: NEWGENE_1561431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236526.2 | XP_006236588.1 | 6525 | |||
XM_006236527.2 | XP_006236589.1 | 2 | 822 | |||
XM_008762939.1 | XP_008761161.1 | 3 | 4555 |

More Information
Other Species Targeted

Gene Ontology Categories:
