siRNA Details
Gene Symbol: Irak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106791.2 | NP_001100261.2 | 2 | 275 | ||
XM_006242193.2 | XP_006242255.1 | 2 | 343 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Irak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106791.2 | NP_001100261.2 | 2 | 275 | ||
XM_006242193.2 | XP_006242255.1 | 2 | 343 |