siRNA Details
Gene Symbol: Zfp280d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108165.2 | NP_001101635.2 | 21 | 2642 | ||
XM_006243356.2 | XP_006243418.1 | 20 | 2609 | |||
XM_006243357.2 | XP_006243419.1 | 21 | 2605 | |||
XM_008766288.1 | XP_008764510.1 | 19 | 2546 |
