siRNA Details
Gene Symbol: Piga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108816.2 | NP_001102286.1 | 5 | 661 | ||
XM_006256870.2 | XP_006256932.1 | 6 | 2911 | |||
XM_006256873.2 | XP_006256935.1 | 6 | 1489 | |||
XM_006256875.2 | XP_006256937.1 | 6 | 3031 | |||
XM_006256876.1 | XP_006256938.1 | 6 | 1545 | |||
XM_008773175.1 | XP_008771397.1 | 5 | 4209 | |||
XM_008773176.1 | XP_008771398.1 | 6 | 3892 | |||
XM_008773177.1 | XP_008771399.1 | 6 | 3480 | |||
XM_008773178.1 | XP_008771400.1 | 6 | 3948 | |||
XM_008773179.1 | XP_008771401.1 | 6 | 2956 | |||
XM_008773180.1 | XP_008771402.1 | 6 | 3012 | |||
XM_008773181.1 | XP_008771403.1 | 6 | 2013 |
