siRNA Details
Gene Symbol: Nlrc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764491.1 | XP_008762713.1 | 6 | 2736 | ||
XM_008764492.1 | XP_008762714.1 | 6 | 2736 | |||
XM_008764493.1 | XP_008762715.1 | 6 | 2736 |
