siRNA Details
Gene Symbol: Hdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014151.2 | NP_001014173.2 | 5 | 564 | ||
XM_006254965.2 | XP_006255027.1 | 3 | 275 | |||
XM_006254966.2 | XP_006255028.1 | 3 | 728 | |||
GenBank mRNA | AY539873.1 | - | 5 | 564 | ||
BC078941.1 | - | 3 | 249 | 7109225 |
