siRNA Details
Gene Symbol: Vom2r5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099462.1 | NP_001092932.1 | 3 | 755 |
Gene Symbol: LOC100909608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758630.1 | XP_008756852.1 | 3 | 851 |
Gene Symbol: LOC102552844 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768523.1 | XP_008766745.1 | 3 | 1179 |
