siRNA Details
Gene Symbol: Fzr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108074.2 | NP_001101544.2 | 3 | 251 | ||
GenBank mRNA | BC162059.1 | - | 3 | 251 | 7115243 |

More Information
Other Species Targeted

Gene Ontology Categories:
