siRNA Details
Gene Symbol: LOC680039 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109388.1 | NP_001102858.1 | 6 | 2239 | ||
XM_006251140.2 | XP_006251202.1 | 6 | 2281 | |||
XM_006251141.2 | XP_006251203.1 | 6 | 2281 | |||
XM_008770360.1 | XP_008768582.1 | 4 | 2002 |
