siRNA Details
Gene Symbol: Pccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017030.2 | NP_058726.2 | 3, 4 | 391 | ||
GenBank mRNA | BC078855.1 | - | 3, 4 | 391 | 7108495 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025835, AK050366, BC055946, BC037082, AF327060, and others.
Gene Ontology Categories:
