siRNA Details
Gene Symbol: Pcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107116.1 | NP_001100586.1 | 4 | 297 | ||
XM_006248762.2 | XP_006248824.1 | 4 | 367 | |||
XM_008768953.1 | XP_008767175.1 | 4 | 352 |
Gene Symbol: Pet100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CK840544.1 | - | 1 | 728 |
