siRNA Details
Gene Symbol: Aven | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107757.1 | NP_001101227.1 | 3 | 249 | ||
XM_006234707.2 | XP_006234769.1 | 4 | 623 | |||
XM_008762093.1 | XP_008760315.1 | 4 | 623 |
