siRNA Details
Gene Symbol: Myoz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106469.1 | NP_001099939.1 | 3, 4 | 346 | ||
XM_006233264.2 | XP_006233326.1 | 4, 5 | 561 | |||
GenBank mRNA | FQ215232.1 | - | 4, 5 | 456 | ||
FQ215285.1 | - | 4, 5 | 486 | |||
FQ215331.1 | - | 4, 5 | 515 | |||
FQ215936.1 | - | 4, 5 | 456 | |||
FQ216537.1 | - | 4, 5 | 517 | |||
FQ216608.1 | - | 4, 5 | 534 |
