siRNA Details
Gene Symbol: Zic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108176.1 | NP_001101646.1 | 3 | 727 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009576, AK078127, AK028305, AK035908.
Gene Ontology Categories:
