siRNA Details
Gene Symbol: Trex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024989.1 | NP_001020160.1 | 2 | 105 | ||
GenBank mRNA | BC091242.1 | - | 2 | 105 | ||
FQ233170.1 | - | 2 | 155 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024989.1 | NP_001020160.1 | 2 | 105 | ||
GenBank mRNA | BC091242.1 | - | 2 | 105 | ||
FQ233170.1 | - | 2 | 155 |