siRNA Details
Gene Symbol: Mtus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100989.1 | NP_001094459.1 | 7 | 790 | ||
XM_008768993.1 | XP_008767215.1 | 11 | 1695 | |||
XM_008768994.1 | XP_008767216.1 | 11 | 1549 | |||
GenBank mRNA | BC127538.1 | - | 7 | 790 |
