siRNA Details
Gene Symbol: Lypd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110295.1 | NP_001103765.1 | 4 | 584 | ||
XM_008758982.1 | XP_008757204.1 | 4 | 389 | |||
XM_008758983.1 | XP_008757205.1 | 4 | 534 | |||
XM_008758984.1 | XP_008757206.1 | 4 | 384 |
Gene Symbol: LOC100910410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750649.3 | XP_003750697.1 | 4 | 742 |
