siRNA Details
Gene Symbol: Hacd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101001.1 | NP_001094471.1 | 2 | 314 | ||
XM_006254292.2 | XP_006254354.1 | 1 | 987 | |||
XM_006254293.2 | XP_006254355.1 | 1 | 988 | |||
GenBank mRNA | AM260199.1 | - | 2 | 314 |

More Information
Other Species Targeted

Gene Ontology Categories:
