siRNA Details
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108547.2 | NP_001102017.2 | 23 | 2596 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108547.2 | NP_001102017.2 | 23 | 2596 |