siRNA Details
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108547.2 | NP_001102017.2 | 22 | 2523 | ||
GenBank mRNA | BC168962.1 | - | 5 | 497 |

More Information
Other Species Targeted

Gene Ontology Categories:
